Learn More
IBA Lifesciences CD4 Isolation Kit for FABian™, Mouse
Product Code. 16242931 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit
10 Kits
Unit Size:
1 set
10 sets
Product Code. 16242931
Brand: IBA Lifesciences 66501001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads
  • The kit is used for the selection of CD4+ T cells from mouse spleen in conjunction with the FABian&trade system.
  • It includes CD4 Fab-Strep (mouse), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian&trade run to isolate up to 3x10^7 target cells.
  • Specifications
    Product Type Non-magnetic Cell Isolation Kit
    Content And Storage 2°C to 8°C
    Donor Source Mouse Spleen
    For Use With (Application) Isolation of label-free cells
    Sufficient For 3x107 target cells
    Quantity 1 Kit