Learn More
CD4 Antibody (CT-4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-28267
Product Code. 18557711
  • 632.00€ / 250mg
Description
CD4 Monoclonal antibody specifically detects CD4 in Avian, Chicken, Turkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
FITC
CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Chicken thymocytes and Ig-negative blood leukocytes
0.25 mg
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
920
Store at 4°C. Do not freeze.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
CT-4
PBS
Mouse
Protein A or G purified
RUO
Primary
Avian, Chicken, Turkey