missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (CT-4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28266
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Avian, Chicken, Turkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Chicken thymocytes and Ig-negative blood leukocytes | |
| 0.5 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CT-4 | |
| Unconjugated | |
| 0.1M BBS (pH 8.2) | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Avian, Chicken, Turkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction