missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Mouse anti-Human, Mouse, Clone: 3C6G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60006-1-IG-150UL
This item is not returnable.
View return policy
Description
CD38 (NAD+ glycohydrolase) is a type II transmembrane glycoprotein able to induce activation, proliferation and differentiation of mature lymphocytes and mediate apoptosis of myeloid and lymphoid progenitor cells. CD38 functions as a multi-catalytic ectoenzyme serving as ADP-ribosyl cyclase, cyclic ADP-ribose hydrolase and possibly NAD+ glycohydrolase or as a cell surface receptor. Antibodies to CD38 are useful in subtyping of lymphomas and leukemias, detection of plasma cells (i.e. identification of myelomas), and as a marker for activated B and T cells. CD38 participates in cell adhesion, signal transduction and calcium signaling. Further, CD38 is expressed at high levels in the pancreas, liver, kidney, maligt lymphoma and neuroblastoma. Diseases associated with CD38 dysfunction include chronic lymphocytic leukemia and Richter′s Syndrome.Specifications
| CD38 | |
| Monoclonal | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28907, P56528 | |
| CD38 | |
| CD38 Fusion Protein Ag0487 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 3C6G4 | |
| Unconjugated | |
| CD38 | |
| ADP ribosyl cyclase 1, cADPr hydrolase 1, CD38, CD38 molecule, Cyclic ADP ribose hydrolase 1, T10 | |
| Mouse | |
| Protein G | |
| RUO | |
| 12494, 952 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction