missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (8C12), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-81049MFV450
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD35 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 8C12 | |
| Flow Cytometry | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Purified murine CR1 protein. | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction