missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CD34 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HPCA1/4393R |
| Conjugate | DyLight 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to CD34 residues within aa285-385 of CD34 was used as an immunogen |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?