missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (CL14579) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26871FR
This item is not returnable.
View return policy
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Immunohistochemistry
Specifications
| CD34 | |
| Monoclonal | |
| DyLight 680 | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| This antibody was developed using a recombinant protein derived from P28906, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| 947 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| CL14579 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction