Learn More
CD34 Antibody (CL14579) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26871DL594
Product Code. 30290776
  • 737.00€ / 0.10mL
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD34
Monoclonal
DyLight 594
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
This antibody was developed using a recombinant protein derived from P28906, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
947
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry
CL14579
50mM Sodium Borate
Mouse
Protein A purified