missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD326 (EpCAM) Mouse anti-Human , APC, Clone: 9C4, BD Biosciences™
Description
CD326 (EpCAM) Monoclonal antibody specifically detects CD326 (EpCAM) in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD326 (EpCAM) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 9C4 |
| Conjugate | APC |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | EGP-2; EGP314; EGP40; EPCAM; ESA; GA733-2; KSA; M4S1; MIC18; MK-1; TACSTD1; TROP1; adenocarcinoma-associated antigen |
| Host Species | Mouse |
| Immunogen | Human breast carcinoma Cell Line |
| Quantity | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?