missing translation for 'onlineSavingsMsg'
Learn More

CD31/PECAM-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

302.00€ - 518.70€

Specifications

Antigen CD31/PECAM-1
Dilution Immunohistochemistry-Paraffin 1:100 - 1:500
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18238050
View Documents
Novus Biologicals
NBP2-54655
100 μL
ONLINE EXCLUSIVE PRICE
549.00€
518.70€ / 100µL
Save 30.30€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18610857
View Documents
Novus Biologicals
NBP2-54655-25ul
25 μL
302.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD31/PECAM-1
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
5175
This CD31/PECAM-1 Antibody was developed against a recombinant Protein corresponding to amino acids: APPANFTIQKEDTIVSQTQDFTKIASKSDSGTYICTAGIDKVVKKSNTVQIVVCEMLSQPRISYDAQFEVIKGQTIEVRCESISGTLPISYQLLKTSKVLENSTKNSNDPAVFKDNPTEDVEYQCVADNCHSHAKMLSEVL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry-Paraffin 1:100 - 1:500
Polyclonal
Rabbit
Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
PECAM1
IgG
Affinity Purified
82.5 kDa
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.