missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LG (Nepmucin) Rat anti-Mouse, BV421, Clone: ZAQ2, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 752372
This item is not returnable.
View return policy
Description
CD300LG (Nepmucin) Monoclonal antibody specifically detects CD300LG (Nepmucin) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD300LG (Nepmucin) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms CD300LG (Nepmucin) BV421 ZAQ2 50 μg | |
| Mouse Nepmucin extracellular domain | |
| RUO | |
| CD300LG (Nepmucin) | |
| Refrigerate (2 to 8°C) | |
| Mouse |
| Flow Cytometry | |
| ZAQ2 | |
| Brilliant Violet 421 | |
| Rat | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction