missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD300c Monoclonal antibody specifically detects CD300c in Human samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Clone | 103 |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CD300c (Accession#: NP_006669.1; Met1-Arg183). |
| Regulatory Status | RUO |
| Gene ID (Entrez) | 10871 |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?