missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Rat anti-Mouse, BV480, Clone: 17A2, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 565642
This item is not returnable.
View return policy
Description
CD3 Monoclonal antibody specifically detects CD3 in Mouse samples. It is validated for Immunofluorescence.
Specifications
| CD3 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Immunofluorescence | |
| 17A2 | |
| Brilliant Violet 480 | |
| CD3; CD3 epsilon; Cd3e; CD3ε; T3e | |
| γδ TCR-positive T-T hybridoma D1 | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV480 under optimum conditions and unconjugated antibody and free BD Horizon BV480 were removed. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction