missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 epsilon Antibody (UCHT1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2726SS
This item is not returnable.
View return policy
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human, Porcine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunodiffusion, Dual RNAscope ISH-IHC.
Specifications
| CD3 epsilon | |
| Monoclonal | |
| 1.47 mg/mL | |
| Western Blot, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunodiffusion, Dual RNAscope ISH-IHC | |
| CD3E | |
| Human infant thymocytes and Sezary cells. | |
| 0.025 mg | |
| Cytokine Research, Mesenchymal Stem Cell Markers, Stem Cell Lines, Stem Cell Markers | |
| 916 | |
| Human | |
| Purified |
| Western Blot | |
| UCHT1 | |
| Unconjugated | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Detects the non-glycosylated epsilon chain of CD3 (MW 20kD) which is closely associated on the lymphocyte cell surface with the T cell antigen receptor (TCR). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction