missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Mouse anti-Human, NA/LE, Clone: CD28.2, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 567117
This item is not returnable.
View return policy
Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow cytometry, Activation
Specifications
| CD28 | |
| Low Endotoxin/Azide-free (LE/AF) | |
| 1.0 mg/mL | |
| CD28 antigen; T44; Tp44; TP44 | |
| Human CD28 Transfected Cell Line | |
| 2.0 mg | |
| Primary | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions. |
| Flow Cytometry | |
| CD28.2 | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, sterile filtered(0.2μm pore size membrane). Endotoxin level is ≤0.1 EU/μg (≤0.01 ng/μg) of protein as determined by the LAL assay. | |
| Mouse | |
| Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction