Description
- The kit is used for the selection of CD28+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
- It includes CD28 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
- Each kit is sufficient for one FABian™ run to isolate cells from up to 9mL whole blood or up to 6.25mL buffy coat.
Specifications
Specifications
| Product Type | Non-magnetic Cell Isolation Kit |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Isolation of label-free cells |
| Quantity | 10 Kits, Human |