Learn More
IBA Lifesciences CD28 Isolation Kit for FABian™
Product Code. 15889774 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit, Human
10 Kits, Human
Unit Size:
1 set
10 sets
Product Code. 15889774
Brand: IBA Lifesciences 66014001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
IBA Lifesciences™ CD28 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
  • The kit is used for the selection of CD28+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD28 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from up to 9mL whole blood or up to 6.25mL buffy coat.
Specifications
Product Type Non-magnetic Cell Isolation Kit
Content And Storage 2°C to 8°C
For Use With (Application) Isolation of label-free cells
Quantity 1 Kit, Human