missing translation for 'onlineSavingsMsg'
Learn More

CD279 (PD-1) Mouse anti-Human , PE-CF594, Clone: MIH4, BD Biosciences™

Mouse Monoclonal Antibody

156.57€ - 410.33€

Specifications

Antigen CD279 (PD-1)
Clone MIH4
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-CF594
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17217654
View Documents
BD Biosciences
566846
25 Tests
ONLINE EXCLUSIVE PRICE
157.00€
156.57€ / 25 tests
Save 0.43€
0.27% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17207654
View Documents
BD Biosciences
566845
100 Tests
ONLINE EXCLUSIVE PRICE
411.00€
410.33€ / 100 tests
Save 0.67€
0.16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD279 (PD-1)
Flow Cytometry
PE-CF594
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human PD-1 Transfected Cell Line
Primary
MIH4
Monoclonal
Mouse
Human
PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.