missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Mouse anti-Human, Brilliant Ultraviolet 737, Clone: EH12.1 (also known as EH12), BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612791
Additional Details : Weight : 0.00060kg
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.Specifications
CD279 (PD-1) | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2 | |
Human PD-1 Recombinant Protein | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
EH12.1 (also known as EH12) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |