missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse anti-Human, Alexa Fluor 647, Clone: O323, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 567026
Additional Details : Weight : 0.00720kg
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow cytometrySpecifications
CD27 | |
Monoclonal | |
Alexa Fluor 647 | |
CD27 antigen; TNFRSF7; S152; T14; Tp55 | |
Not Reported | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
O323 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |