missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD267 (TACI) Rat, anti-Mouse, APC, Clone: 8F10, STEMCELL Technologies
Description
- The 8F10 antibody reacts with mouse CD267 (transmembrane activator and CAML interactor, or TACI), an ∼30 kDa type III transmembrane protein of the TNF receptor superfamily that is expressed primarily on the surface of maturing subsets of splenic B cells, including transition type 2 (T2) and marginal zone (MZ) B cells, as well as on plasma cells and plasmablasts
- Expression of CD267 is upregulated on B cells exposed to LPS both in vitro and in vivo
- CD267 plays important roles in humoral immunity by regulating B cell survival, proliferation, and maturation through its interaction with the ligands APRIL and BAFF
- Ligand binding stimulates activation of several transcription factors, including NF-kappa-B, and has been shown to induce either proliferation or apoptosis, suggesting a dual role in the regulation of B cells
- CD267 also appears to have an essential function in immunoglobulin isotype switching and B cell differentiation in response to thymus-independent antigens
Specifications
Specifications
| Antigen | CD267 (TACI) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 8F10 |
| Conjugate | APC |
| Host Species | Rat |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Target Species | Mouse |
| Isotype | IgG2a κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?