missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2R alpha Rat anti-Mouse, Rat, Clone: AP-MAB0710, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB110-97867-0.1mg
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with recombinant mouse CD25 protein. | |
| 0.1 mg | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
| 3559 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| AP-MAB0710 | |
| Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:300, CyTOF-ready | |
| P01590 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.4 mL sterilized PBS to a final concentration of 0.5 mg/mL. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction