missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2R alpha Antibody (1B5D12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00€ - 603.00€
Specifications
| Antigen | CD25/IL-2R alpha |
|---|---|
| Clone | 1B5D12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613756
|
Novus Biologicals
NBP2-37349-0.025ml |
0.025 mL |
196.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18166417
|
Novus Biologicals
NBP2-37349 |
0.1 mL |
603.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CD25/IL-2R alpha | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Monkey, Primate | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| IL2RA | |
| IgG1 | |
| Affinity Purified |
| 1B5D12 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 3559 | |
| Purified recombinant fragment of human IL2RA (AA: 34-139) expressed in E. Coli. | |
| Primary | |
| 30.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title