Learn More
CD24 Antibody (SN3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-64861-0.025MG
This item is not returnable.
View return policy
Description
CD24 Monoclonal specifically detects CD24 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD24 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1/50-1/100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500 | |
| P25063 | |
| CD24 | |
| Glycoproteins purified from human NALM-1 cell line | |
| 0.025 mg | |
| Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| 100133941 | |
| Human | |
| Purified |
| Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| SN3 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes the human CD24 cell surface antigen, a heavily glycosylated molecule of 35-45kD. CD24 is expressed by granulocytes and B lymphocytes, and by some activated T cells and T cell malignancies. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |