missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23/Fc epsilon RII Antibody (EBVCS2), Novus Biologicals™
Mouse Monoclonal Antibody
285.00€ - 474.00€
Specifications
| Antigen | CD23/Fc epsilon RII |
|---|---|
| Clone | EBVCS2 |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277494
|
Novus Biologicals
NBP1-43433-0.025MG |
0.025 mg |
285.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18272853
|
Novus Biologicals
NBP1-43433-0.1MG |
0.1 mg |
474.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| CD23/Fc epsilon RII | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Adaptive Immunity, Cancer, Immunology | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 2208 | |
| The immunogen for this antibody was CD23. | |
| Primary | |
| Store at 4C. Do not freeze. |
| EBVCS2 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human | |
| BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor | |
| FCER2 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title