missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Antibody (GB25A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60855
This item is not returnable.
View return policy
Description
CD21 Monoclonal specifically detects CD21 in Bovine, Goat, Bison, Water Buffalo, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD21 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Mouse | |
| Unpurified | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology | |
| 1380 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Functional Assay | |
| GB25A | |
| Unconjugated | |
| Ascites with 0.09% Sodium Azide | |
| CR2 | |
| Goat ConA stimulated mononuclear leukocytes | |
| 0.1 mg | |
| Primary | |
| Bovine, Goat, Bison, Water Buffalo, Sheep | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction