Learn More
CD21 Antibody (CL14552) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26866JF669
Product Code. 30299899
  • 770.00€ / 0.10mL
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD21
Monoclonal
Janelia Fluor 669
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
This antibody was developed using a synthetic peptide derived from P20023, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology
1380
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot
CL14552
50mM Sodium Borate
Mouse
Protein A purified