missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Antibody (544411) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB49091I
This item is not returnable.
View return policy
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Immunohistochemistry
Specifications
| CD21 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD21, Ile21-Arg971, Accession # P20023 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology | |
| 1380 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| 544411 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction