Learn More
CD209 / DC-SIGN Mouse, Clone: C209/1781, enQuireBio™
Product Code. 15900168
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15900168 100 μg 100µg
15990158 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15900168 Supplier EnquireBio Supplier No. 30835MSM1P1
Mouse Monoclonal Antibody
Specifications
Antigen CD209 / DC-SIGN
Applications Immunohistochemistry
Cellular Localization Secreted and Cell Surface
Classification Monoclonal
Clone C209/1781
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% Azide
Gene Accession No. NP_001138371
Gene Symbols CD209
Host Species Mouse
Immunogen Recombinant human CD209 protein fragment
Molecular Weight of Antigen 45kDa
Positive Control THP-1 or HeLa Cells. Small Intestine, Uterus or Lymph Node.
Purification Method Protein A/G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30835
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG2b κ