missing translation for 'onlineSavingsMsg'
Learn More

CD197 (CCR7) Mouse anti-Human, Brilliant Violet 711, Clone: 150503, Secondary Antibody, BD Horizon™

Product Code. 15875179
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15875179 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15875179 Supplier BD Biosciences Supplier No. 566602
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD197 (CCR7) Monoclonal antibody specifically detects CD197 (CCR7) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD197 (CCR7)
Applications Flow Cytometry
Classification Monoclonal
Clone 150503
Conjugate Brilliant Violet 711
Host Species Mouse
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity CD197 (CCR7)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.