Description
- The kit is used for the selection of CD19+ cells from whole blood or buffy coat in conjunction with the FABian™ system
- It includes CD19 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe
- Each kit is sufficient for one FABian™ run to isolate cells from 9 ml whole blood or 6.4 mL buffy coat
Specifications
Specifications
| Product Type | Cell Isolation Kit |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Selection of CD19+ Cells From Whole Blood or Buffy Coat in Conjunction with The FABian™ System |
| Quantity | 10 Kits |
| Format | 10 Columns |