Learn More
CD184 Rat anti-Mouse, Oligo, Clone: 2B11/CXCR4, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand:  BD Biosciences 940151
Product Code. 17203215
  • 439.00€ / 25 tests
Description
CD184 Monoclonal antibody specifically detects CD184 in Mouse samples. It is validated for Single Cell 3' Sequencing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD184
Monoclonal
Oligo
CXCR4, C-X-C chemokine receptor type 4; Fusin; LESTR; PB-CKR; Sdf1r
GST-NCXCR4 fusion protein
RUO
Mouse
IgG2b κ
Single Cell 3' Sequencing
Clone 2B11/CXCR4
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions.