missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD177 Rat anti-Mouse, Alexa Fluor™ 647, Clone: Y127, BD Pharmingen™
Rat Monoclonal Antibody
Brand: BD Biosciences 566599
This item is not returnable.
View return policy
Description
CD177 Monoclonal antibody specifically detects CD177 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD177 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| Y127 | |
| Alexa Fluor 647 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction