missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161/NK1.1 Mouse anti-Mouse, Clone: PK136, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07532-100ug
This item is not returnable.
View return policy
Description
CD161/NK1.1 Monoclonal specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.
Specifications
| CD161/NK1.1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5-1 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Functional | |
| CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| CD Mouse spleen and bone marrow cells enriched for NK-1+ cells | |
| 100 μg | |
| Innate Immunity, Signal Transduction | |
| 17059 | |
| Store at 4°C. | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Functional Assay | |
| PK136 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human (Negative), Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction