missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Antibody (702228), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7448H
This item is not returnable.
View return policy
Description
CD161 Monoclonal antibody specifically detects CD161 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CD161 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 702228 | |
| Flow Cytometry, CyTOF-ready | |
| CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| BaF3 mouse pro-B cell line transfected with human CD161, Accession # Q12918 | |
| 0.1 mL | |
| Innate Immunity, Signal Transduction | |
| 3820 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction