Learn More
CD161 Antibody (14F1F11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31359-0.025MG
This item is not returnable.
View return policy
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD161 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 5ug/ml, Immunohistochemistry 1:10 - 1:2000, Immunohistochemistry-Paraffin 5 ug/ml | |
| Q12918 | |
| KLRB1 | |
| Amino acids 68-225 of human CD161 were used as the immunogen for this antibody. | |
| 0.025 mg | |
| Signal Transduction | |
| 3820 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 14F1F11 | |
| Unconjugated | |
| PBS with No Preservative | |
| CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The antibody has not been tested in species samples other than human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |