missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Hamster anti-Mouse, BV480, Clone: HL3, BD Horizon™
Hamster Monoclonal Antibody
Brand: BD Biosciences 565627
This item is not returnable.
View return policy
Description
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Immunofluorescence.
Specifications
| CD11c | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Hamster | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Immunofluorescence | |
| HL3 | |
| Brilliant Violet 480 | |
| Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4 | |
| C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV480 under optimum conditions and unconjugated antibody and free BD Horizon BV480 were removed. | |
| IgG1 γ2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction