missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 Rat anti-Mouse, Brilliant Ultraviolet 737, Clone: ACK45, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 741762
This item is not returnable.
View return policy
Description
CD117 Monoclonal antibody specifically detects CD117 in Mouse samples. It is validated for Flow cytometry
Specifications
| CD117 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon Brilliant Ultraviolet 737 under optimal conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| ACK45 | |
| Brilliant Ultraviolet 737 | |
| c-KIT; W; SCFR; Stem Cell Factor Receptor; Sl; Steel Factor Receptor; Ssm | |
| Mouse bone marrow mast cells | |
| RUO | |
| Mouse | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction