missing translation for 'onlineSavingsMsg'
Learn More

CD117/c-kit Antibody (kit2c75) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

225.00€

Specifications

Antigen CD117/c-kit
Clone kit2c75
Concentration 0.5 mg/mL
Dilution Flow Cytometry ≤ 1 μg/106 cells, Immunocytochemistry/Immunofluorescence
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18584643
View Documents
Novus Biologicals
NBP1-28327-0.1mg
0.1 mg
225.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD117/c-kit Monoclonal specifically detects CD117/c-kit in Chicken samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CD117/c-kit
0.5 mg/mL
Flow Cytometry
Unconjugated
Mouse
Cancer, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
KIT
IgG2a κ
Protein A or G purified
Chicken c-Kit
kit2c75
Flow Cytometry ≤ 1 μg/106 cells, Immunocytochemistry/Immunofluorescence
Monoclonal
Purified
RUO
Chicken
3815
Sp/chkit4A4 cells (Sp2/0 cells transfected with chicken c-Kit cDNA)
Primary
Store at 4°C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.