missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD10 Mouse anti-Human APC-R700, Clone: HI10a, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 659120
This item is not returnable.
View return policy
Description
CD10 Monoclonal antibody specifically detects CD10 in Human samples. It is validated for Flow Cytometry
Specifications
| CD10 | |
| Monoclonal | |
| 12.5 μg/mL | |
| Phosphate buffered saline (PBS) containing BSA and ProClin. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human |
| Flow Cytometry | |
| Clone HI10a | |
| APC-R700 | |
| MME; CALLA; EPN; NEP; neprilysin; SFE; atriopeptidase; enkephalinase | |
| Acute CALLA Leukemia Blast Cells | |
| RUO | |
| 12481 | |
| IgG1 κ |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- CD10 APC-R700 (HI10a) RUO
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction