missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD-M6PR Antibody (PSH01-06), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32555
This item is not returnable.
View return policy
Description
CD-M6PR Monoclonal antibody specifically detects CD-M6PR in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CD-M6PR | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
| Recombinant protein within human CD-M6PR aa 1-200/ 277. (Uniprot: P20645) | |
| 100 μL | |
| Immunology | |
| 4074 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| PSH01-06 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction