missing translation for 'onlineSavingsMsg'
Learn More

CCT4 Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene

Product Code. 17514404
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17514404 30 μL 30µL
17594394 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17514404 Supplier Origene Technologies Supplier No. TA810867S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCT4 Monoclonal antibody specifically detects CCT4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CCT4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Hnrnph1
Gene Accession No. P31943
Gene Alias 2610204B21Rik; A45; C78323; CCT4; cct4 protein; Cctd; CCT-delta; chaperonin containing t-complex polypeptide 1, delta subunit; chaperonin containing TCP1 subunit 4; chaperonin containing Tcp1, subunit 4 (delta); chaperonin subunit 4 (delta); SRB; Stimulator of TAR RNA-binding; T complex protein 1, delta; T-complex protein 1 subunit delta; TCP-1 delta; TCP-1-delta; Unknown (protein for MGC:134500); zgc:65789; zgc:77192
Host Species Mouse
Immunogen Full length human recombinant protein of human HNRNPH1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3187
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.