missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR8 Antibody (10A9), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-26689-100ul
This item is not returnable.
View return policy
Description
CCR8 Monoclonal antibody specifically detects CCR8 in Human samples. It is validated for Flow Cytometry
Specifications
| CCR8 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 10A9 | |
| Flow Cytometry | |
| CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1 | |
| Recombinant Human CCR8 protein [UniProt P51685] | |
| 100 μL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| 1237 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction