Learn More
CCR6 Antibody (1011306), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1951C
Product Code. 18644753
  • 559.00€ / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
CCR6 Monoclonal antibody specifically detects CCR6 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCR6
Monoclonal
PerCP
PBS
Mouse
Protein A or G purified from ascites
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
1011306
Flow Cytometry, CyTOF-ready
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
NS0 mouse myeloma cell line transfected with human CCR6, Met1-Met374, Accession # P51684
0.1 mL
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
1235
Store at 4°C in the dark.