missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS1441
This item is not returnable.
View return policy
Description
CCR11 Polyclonal antibody specifically detects CCR11 in Human, Mouse, Rat, Porcine, Bat, Canine, Equine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CCR11 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 24 μg/mL | |
| CC chemokine receptor-like 1, C-C CKR-11, CCBP2CCX CKR, CCR11CCR10, chemokine (C-C motif) receptor-like 1, orphan seven-transmembrane receptor, chemokine related, PPR1cc motif, receptor-like protein 1, VSHK1CKR-11 | |
| Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human CCR11. | |
| 0.05 mL | |
| GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 51554 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction