missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCR1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149108
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Chemokines play important roles in inflammation and are critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines. One of the CCR receptors, CCR1, is expressed on neutrophils, monocytes, lymphocytes, and eosinophils and binds the leukocyte chemoattractant and hemopoiesis regulator macrophage-inflammatory protein (MIP-1 ), eotaxin, as well as several other related chemokines. Mice lacking the chemokine receptor CCR1 have defects in neutrophil trafficking and proliferation.
Specifications
| CCR1 | |
| Polyclonal | |
| Unconjugated | |
| CCR1 | |
| C-C chemokine receptor type 1; C-C CKR-1; C-C motif chemokine receptor 1; CC-CKR-1; Ccr1; CCR-1; CD191; chemokine (C-C motif) receptor 1; chemokine (C-C) receptor 1; CKR1; CKR-1; CMKBR1; CMKR1; HM145; LD78 receptor; macrophage inflammatory protein 1-alpha receptor; macrophage inflammatory protein-1 alpha receptor; MIP-1 alpha R; MIP-1 alphaR; MIP-1alpha-R; MIP1aR; Mip-1a-R; RANTES receptor; RANTES-R; SCYAR1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 12768 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| P51675 | |
| CCR1 | |
| E. coli-derived recombinant mouse CCR1, Met1-Ala34, Asp92-Lys107, Phe172-Arg197, Gln265-Leu281. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction