missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL4/MIP-1 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77897
This item is not returnable.
View return policy
Description
CCL4/MIP-1 beta Polyclonal specifically detects CCL4/MIP-1 beta in Porcine samples. It is validated for Western Blot, ELISA.
Specifications
| CCL4/MIP-1 beta | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP-1 beta/CCL4, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2 | |
| Rabbit | |
| 10.21 kDa | |
| 0.1 mg | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6351 | |
| Pig | |
| Purified |
| Western Blot, ELISA | |
| 1 mg/ml | |
| Western Blot 1:500, ELISA 1:5000 | |
| Q8NHW4 | |
| CCL4 | |
| This protein-A purified CCL4/MIP-1 beta Antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant protein raised in yeast, corresponding to the 69 amino acids of the mature swine CCL4/MIP-1 beta protein. (Uniprot: Q711P4) | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody is specific for swine CCL4 protein. A BLAST analysis was used to suggest cross-reactivity with CCL4 from swine sources based on 100% homology with the immunogen. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction