missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL21/6Ckine Monoclonal antibody specifically detects CCL21/6Ckine in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | CCL21/6Ckine |
| Applications | Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) |
| Classification | Monoclonal |
| Clone | 59106 |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine |
| Host Species | Rat |
| Immunogen | E. coli-derived recombinant mouse CCL21/6Ckine, Ser24-Gly133, Accession # P84444 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?