missing translation for 'onlineSavingsMsg'
Learn More

CCHCR1 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16831425 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16831425 20 μL 20µL
16821425 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16831425 Supplier Proteintech Supplier No. 218691AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCHCR1 may be a regulator of keratinocyte proliferation or differentiation.

Specifications

Antigen CCHCR1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCHCR1
Gene Accession No. Q8TD31
Gene Alias C6orf18, CCHCR1, HCR, Pg8, Putative gene 8 protein, SBP
Gene Symbols CCHCR1
Host Species Rabbit
Immunogen CCHCR1 Fusion Protein Ag16141
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54535
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.