missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC83 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 462.00€
Specifications
| Antigen | CCDC83 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CCDC83 Polyclonal antibody specifically detects CCDC83 in Human samples. It is validated for Western BlotSpecifications
| CCDC83 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS with 50% glycerol, pH7.3. | |
| 220047 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| coiled-coil domain containing 83, coiled-coil domain-containing protein 83, FLJ42119, MGC34732 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CCDC83 (NP_001273088.1). MENSGKANKKDTHDGPPKEIKLPTSEALLDYQCQIKEDAVEQFMFQIKTLRKKNQKYHERNSRLKEEQIWHIRHLLKELSEEKAEGLPVVTREDVEEAMKEKWKFERDQEKNLRDMRMQISNAEKLFLEKLSEKEYWEEYKNVGSERHAKLITSLQNDINTVKENAEKMSEHYKITLEDTRKKIIKETLLQLDQKKEWATQNAVKLIDKGSYLEIWENDWLKKEVAIHRKEVEELKNAIHELEAENLVLIDQLSNCRLVDLKIPRRLYLTQAAGLEVPPEEMSLELPETHIEEKSELQPT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title